ENA447957064 Enamine

4-chloro-1H-1,3-benzodiazol-2-amine hydrochloride

  • MDL number MFCD28397492
  • Cas Number : 1803612-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207828

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207828
Availability details for
Quantity
  • Availability
  • Add to Cart