ENA447957067 Enamine

3-[5-(thiophen-2-yl)-1,3,4-oxadiazol-2-yl]morpholine hydrochloride

  • MDL number MFCD28397496
  • Cas Number : 1803604-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207875

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207875
Availability details for
Quantity
  • Availability
  • Add to Cart