ENA447957069 Enamine

3-(5-phenyl-1,3,4-oxadiazol-2-yl)morpholine hydrochloride

  • MDL number MFCD28397499
  • Cas Number : 1803601-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207879
Availability details for
Quantity
  • Availability
  • Add to Cart