ENA447957071 Enamine

4-ethoxy-3,3-dimethylpiperidine

  • MDL number MFCD28397502
  • Cas Number : 1803604-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207890
Availability details for
Quantity
  • Availability
  • Add to Cart