ENA447957072 Enamine

{1-[(1,2,4-oxadiazol-3-yl)methyl]piperidin-3-yl}methanamine dihydrochloride

  • MDL number MFCD28397503
  • Cas Number : 1803566-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207917
Availability details for
Quantity
  • Availability
  • Add to Cart