ENA447957073 Enamine

2-(4-chloropyridin-3-yl)acetic acid hydrochloride

  • MDL number MFCD28397504
  • Cas Number : 1803562-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207931
Availability details for
Quantity
  • Availability
  • Add to Cart