ENA447957082 Enamine

sodium 2-(4-methyl-1,3-thiazol-2-yl)propanoate

  • MDL number MFCD28397514
  • Cas Number : 1803593-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207986

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207986
Availability details for
Quantity
  • Availability
  • Add to Cart