ENA447957083 Enamine

2-(2-amino-3-phenylpropanamido)pent-4-enoic acid hydrochloride

  • MDL number MFCD28397516

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207993
Availability details for
Quantity
  • Availability
  • Add to Cart