ENA447957087 Enamine

2-(1,3-dioxolan-2-yl)-5-iodo-1,3-thiazole

  • MDL number MFCD28397520
  • Cas Number : 1225029-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208046

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208046
Availability details for
Quantity
  • Availability
  • Add to Cart