ENA447957091 Enamine

2-[5-(4-methoxypiperidin-4-yl)-1,2,4-oxadiazol-3-yl]pyridine

  • MDL number MFCD28397524
  • Cas Number : 1803565-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208060
Availability details for
Quantity
  • Availability
  • Add to Cart