ENA447957106 Enamine

5-amino-3-(trifluoromethyl)pentanoic acid hydrochloride

  • MDL number MFCD28397531
  • Cas Number : 1803565-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208186
Availability details for
Quantity
  • Availability
  • Add to Cart