ENA447957107 Enamine

methyl 2-amino-2-(4-chlorophenyl)butanoate hydrochloride

  • MDL number MFCD28397532
  • Cas Number : 1803585-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208187
Availability details for
Quantity
  • Availability
  • Add to Cart