ENA447957111 Enamine

3,5-dimethyl-1-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}-1H-pyrazole

  • MDL number MFCD28397537
  • Cas Number : 2246678-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208215
Availability details for
Quantity
  • Availability
  • Add to Cart