ENA447957113 Enamine

ethyl 3-(4-chlorophenyl)-1-methyl-1H-1,2,4-triazole-5-carboxylate

  • MDL number MFCD28397540
  • Cas Number : 1803589-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208237

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208237
Availability details for
Quantity
  • Availability
  • Add to Cart