ENA447957115 Enamine

1-(3-aminopyrrolidin-1-yl)-2-phenylethan-1-one

  • MDL number MFCD16667198
  • Cas Number : 727720-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205951
Availability details for
Quantity
  • Availability
  • Add to Cart