ENA447957119 Enamine

2-(chloromethyl)-5-(trifluoromethyl)pyridine

  • MDL number MFCD10697590
  • Cas Number : 128790-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205959
Availability details for
Quantity
  • Availability
  • Add to Cart