ENA447957124 Enamine

ethyl 5-(4-chlorophenyl)-1H-1,2,4-triazole-3-carboxylate

  • MDL number MFCD19141565
  • Cas Number : 62230-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205283
Availability details for
Quantity
  • Availability
  • Add to Cart