ENA447957125 Enamine

2,2-dichloro-N-(4-chlorophenyl)-3-oxo-3-phenylpropanamide

  • MDL number MFCD28383950
  • Cas Number : 19359-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205980

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205980
Availability details for
Quantity
  • Availability
  • Add to Cart