ENA447957134 Enamine

N-(2-methoxyethyl)-3-nitrobenzamide

  • MDL number MFCD00751342
  • Cas Number : 167834-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205294
Availability details for
Quantity
  • Availability
  • Add to Cart