ENA447957138 Enamine

3-amino-N-(2-methoxyethyl)benzamide

  • MDL number MFCD09047611
  • Cas Number : 167837-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205295
Availability details for
Quantity
  • Availability
  • Add to Cart