ENA447957141 Enamine

imidazo[2,1-b][1,3]thiazole-3-carboxylic acid

  • MDL number MFCD21909963
  • Cas Number : 479028-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206060
Availability details for
Quantity
  • Availability
  • Add to Cart