ENA447957143 Enamine

2,2,2-trifluoro-1-(2-methylindolizin-3-yl)ethan-1-one

  • MDL number MFCD02707620
  • Cas Number : 127532-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206067
Availability details for
Quantity
  • Availability
  • Add to Cart