ENA447957148 Enamine

2-[(5-bromo-2-nitrophenyl)(methyl)amino]acetic acid

  • MDL number MFCD21296038
  • Cas Number : 305790-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206076

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206076
Availability details for
Quantity
  • Availability
  • Add to Cart