ENA447957149 Enamine

6-bromo-4-methyl-1,2,3,4-tetrahydroquinoxalin-2-one

  • MDL number MFCD20488827
  • Cas Number : 305790-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206077
Availability details for
Quantity
  • Availability
  • Add to Cart