ENA447957151 Enamine

methyl 4-amino-3-chloro-5-methylbenzoate

  • MDL number MFCD01313708
  • Cas Number : 202146-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-82
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205310

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205310
Availability details for
Quantity
  • Availability
  • Add to Cart