ENA447957153 Enamine

5-chloro-3-(dimethylcarbamoyl)-2-methoxybenzene-1-sulfonyl chloride

  • MDL number MFCD21224442
  • Cas Number : 1482800-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206089
Availability details for
Quantity
  • Availability
  • Add to Cart