ENA447957156 Enamine

(2-amino-4,5-dichlorophenyl)methanol

  • MDL number MFCD20726404
  • Cas Number : 1379313-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206093

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206093
Availability details for
Quantity
  • Availability
  • Add to Cart