ENA447957157 Enamine

methyl 4-bromo-3-chloro-5-methylbenzoate

  • MDL number MFCD23707509
  • Cas Number : 1427440-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205312
Availability details for
Quantity
  • Availability
  • Add to Cart