ENA447957186 Enamine

4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-pyrazole

  • MDL number MFCD22415359
  • Cas Number : 756520-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205325

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205325
Availability details for
Quantity
  • Availability
  • Add to Cart