ENA447957187 Enamine

2-(1-methyl-1H-pyrazol-4-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD25371381
  • Cas Number : 1431962-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206337

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206337
Availability details for
Quantity
  • Availability
  • Add to Cart