ENA447957191 Enamine

3-amino-N-(3-methoxypropyl)-1H-pyrazole-5-carboxamide

  • MDL number MFCD20316303
  • Cas Number : 1347087-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205330
Availability details for
Quantity
  • Availability
  • Add to Cart