ENA447957192 Enamine

2-(4-chlorophenyl)-2-(cyclopropylmethoxy)ethan-1-amine

  • MDL number MFCD16062131
  • Cas Number : 1250007-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206342

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206342
Availability details for
Quantity
  • Availability
  • Add to Cart