ENA447957193 Enamine

1-(5-methyl-4H-1,2,4-triazol-3-yl)ethan-1-amine hydrochloride

  • MDL number MFCD07781071
  • Cas Number : 1803604-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206345

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206345
Availability details for
Quantity
  • Availability
  • Add to Cart