ENA447957196 Enamine

methyl 2-ethyl-1H-pyrrole-3-carboxylate

  • MDL number MFCD12923910
  • Cas Number : 879214-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206360
Availability details for
Quantity
  • Availability
  • Add to Cart