ENA447957201 Enamine

4-methyl-4-phenylazepane

  • MDL number MFCD20690295
  • Cas Number : 933743-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206388
Availability details for
Quantity
  • Availability
  • Add to Cart