ENA447957204 Enamine

2-hydroxy-7,7-dimethyl-1H,4H,6H,7H,8H-[1,3]diazino[1,2-a]pyrimidin-4-one

  • MDL number MFCD23163296
  • Cas Number : 1383777-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205339

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205339
Availability details for
Quantity
  • Availability
  • Add to Cart