ENA447957214 Enamine

N-(4-tert-butyl-1,3-thiazol-2-yl)-3-methylbutanamide

  • MDL number MFCD03744414
  • Cas Number : 626209-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206414
Availability details for
Quantity
  • Availability
  • Add to Cart