ENA447957224 Enamine

tert-butyl N-[4-(3,5-dioxo-1,2,4-triazolidin-4-yl)phenyl]carbamate

  • MDL number MFCD24475867
  • Cas Number : 612838-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206436

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206436
Availability details for
Quantity
  • Availability
  • Add to Cart