ENA447957233 Enamine

4-methoxy-3-sulfamoylbenzamide

  • MDL number MFCD20127178
  • Cas Number : 38500-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206462

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206462
Availability details for
Quantity
  • Availability
  • Add to Cart