ENA447957235 Enamine

3-(4-methoxyphenyl)imidazolidine-2,4-dione

  • MDL number MFCD00450074
  • Cas Number : 62101-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206497
Availability details for
Quantity
  • Availability
  • Add to Cart