ENA447957236 Enamine

{[3,5-bis(trifluoromethyl)phenyl]methyl}(methyl)amine

  • MDL number MFCD22581670
  • Cas Number : 159820-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206498

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206498
Availability details for
Quantity
  • Availability
  • Add to Cart