ENA447957254 Enamine

1-(3-methoxyphenyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD00297151
  • Cas Number : 74205-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206540
Availability details for
Quantity
  • Availability
  • Add to Cart