ENA447957256 Enamine

5-iodobenzene-1,3-diamine

  • MDL number MFCD09036307
  • Cas Number : 111938-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206543
Availability details for
Quantity
  • Availability
  • Add to Cart