ENA447957257 Enamine

2,2-dimethyl-3-phenoxycyclobutan-1-one

  • MDL number MFCD21755626
  • Cas Number : 1520143-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206544
Availability details for
Quantity
  • Availability
  • Add to Cart