ENA447957259 Enamine

7-oxa-1-azaspiro[4.4]nonane hydrochloride

  • MDL number MFCD26960527
  • Cas Number : 1620569-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206548
Availability details for
Quantity
  • Availability
  • Add to Cart