ENA447957265 Enamine

1-{4-[(1-methylpiperidin-4-yl)oxy]phenyl}methanamine

  • MDL number MFCD08699724
  • Cas Number : 937599-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206556
Availability details for
Quantity
  • Availability
  • Add to Cart