ENA447957267 Enamine

decahydronaphthalene-1-carboxylic acid

  • MDL number MFCD20639445
  • Cas Number : 7384-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206559
Availability details for
Quantity
  • Availability
  • Add to Cart