ENA447957268 Enamine

N-[(piperidin-4-yl)methyl]methanesulfonamide

  • MDL number MFCD05022404
  • Cas Number : 166815-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206564
Availability details for
Quantity
  • Availability
  • Add to Cart