ENA447957269 Enamine

[1-(4-aminobenzenesulfonyl)pyrrolidin-2-yl]methanol

  • MDL number MFCD11043095
  • Cas Number : 1306676-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206565

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206565
Availability details for
Quantity
  • Availability
  • Add to Cart