ENA447957274 Enamine

(2R)-2-(cyclohexylmethanesulfonamido)propanoic acid

  • MDL number MFCD16050708
  • Cas Number : 1308979-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206570

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206570
Availability details for
Quantity
  • Availability
  • Add to Cart